North by Honeywell 75SCP100L-12 Cartridge for Full-Face and Half-Mask Respirators, Defender with P100 filter
Representative image only.
North by Honeywell Mfr # 75SCP100L – Item # EW-81606-62
$36.50USD / Pkg of 2
BUY MORE & SAVE
  • 1-17$36.50USD / Pkg of 2
  • 18+$32.85USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • RatingP100
  • NIOSH ApprovalCertain organic vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Fluoride, Ammonia/Methylamine, Hydrogen Sulfide (escape only), Formaldehyde
  • ComplianceNIOSH
  • ColorPink, Orange
  • DescriptionCartridge for Full-Face and Half-Mask Respirators, Defender with P100 filter
More About this Item
Stay safe with replacement cartridges.
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
3 Questions | 3 Answers
Displaying questions 1-3
  • Q:
    do these filters work for the use of spray paint?
    Asked on 4/30/2011 by PJ from UK
    1 answer
    • STAFF EXPERT
      A:
      Yes, a P100 filter is sufficient for protection from spray paint.
      Answered on 5/2/2011 by Jack from Technical Support
  • Q:
    Will this filter protect workers from crystalline silica dust?
    Asked on 11/7/2010 by Suzy from Alberta, Canada
    1 answer
    • STAFF EXPERT
      A:
      The concentration level has to be between 0-.5mg - 1.0mg for these P100 filters to work.
      Answered on 11/9/2010 by Mike from Tech Support
  • Q:
    How many times (or time) each filter can be used?. I want to know how many cartridges for one year I need!
    Asked on 5/7/2010 by Tcha from mexico city
    1 answer
    • STAFF EXPERT
      A:
      The lifetime of the filters will depend on your application including how often and long they are used, what they are filtering and how concentrated it is, etc. Please contact our technical application specialists for additional assistance (contact information can be found on the product web page).
      Answered on 5/11/2010 by David from Tech Support
Displaying questions 1-3
Do you have a question about this product? 