North by Honeywell 11390044 Replacement face shield only, Propionate
Representative image only.
North by Honeywell Mfr # 11390044 – Item # EW-81690-10
Protect yourself against liquid splashes, sparks, and impacts
PRODUCT OPTIONS
$12.50USD / Each
BUY MORE & SAVE
  • 1-9$12.50USD / Each
  • 10+$11.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorClear
  • Lens ColorClear
  • ComplianceANSI Z87.1; CSA Z94.3
  • DescriptionReplacement face shield only, Propionate
More About this Item
SnapLok™Head Gear features adjustable ratchet suspension and tension knob for precise sizing and visor positioning. Splash drip edge redirects splashing or spraying fluids. Sweatband absorbs perspiration. Can be used with earmuffs.
Universal Shields are extra wide for excellent protection. Lightweight construction provides comfort during lengthy work periods. Shields attach easily and securely to head gear—no snaps, posts, or levers.
Model 81690-10 is heat-resistant to temperatures up to 220°>F; model 81690-20 withstands temperatures up to 280°>F. Order shields separately.
Compliance: ANSI Z87.1, CSA Z94.3