This item is now discontinued. We recommend the following as suggested replacements: EW-86232-28
Microflex DF850-XL DermaFree Vinyl Gloves, X-large, box of 100
Representative image only.
Microflex Mfr # DF850-XL – Item # EW-86232-63
Ideal for food processing or light industrial use
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVinyl
  • Thickness (mil)6
  • SizeX-Large
  • Length (in)9
  • Length (cm)23
  • ColorClear
  • SterileNo
  • DescriptionDermaFree Vinyl Gloves, X-large, box of 100
More About this Item
These 100% synthetic vinyl gloves alleviate the potential for adverse reactions from natural rubber proteins. Gloves are thicker for added tear resistance without sacrificing tactile sensitivity. Gloves are powder-free to limit contamination. Lightly beaded cuffs ensure hassle-free removal after use. Ambidextrous.