Sellstrom 80231 Autoclavable Safety Goggles, Top vented
Representative image only.
Sellstrom Mfr # 80231 – Item # EW-86281-02
Sterilize in an autoclave and with gamma radiation for cleanroom environments
$15.90USD / Each
BUY MORE & SAVE
  • 1-9$15.90USD / Each
  • 10+$14.31USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorWhite
  • Lens ColorClear
  • ComplianceANSI Z87.1 (High Impact); cUL-certified to meet CAN/CSA-Z94.3
  • DescriptionAutoclavable Safety Goggles, Top vented
More About this Item
Created for aseptic and cleanroom environments, these autoclavable goggles feature UV-absorbing polycarbonate lenses and a comfortable soft body that easily fits over prescription eyewear.
Compliance: ANSI Z87.1+ (High Impact), CUL certified to meet CAN/CSA-Z94.3