This item is now discontinued. Please browse similar items
SPC810 Boom, 8 inch x 10 foot
Representative image only.
SPC Mfr # SPC810 – Item # EW-86338-02
  • Soak up spills on land or water!
SPECS & DESCRIPTION
Specifications & Description
  • Absorption71 gal./cs
  • Width (in)8
  • Length (ft)10
  • DescriptionBoom, 8 inch x 10 foot
Key Features
  • Soak up spills on land or water!
More About this Item
SPC sorbents are ideal for cleaning up or removing oil from land or water. A single pound of SPC sorbent equals 25 pounds of clay. Each sorbent absorbs up to 25 times its own weight in petroleum products. SPC sorbents are also effective for soaking up petroleum based solvents, paints, vegetable oils, and many other non-water soluble spills. Oil sorbents will not absorb water, will float indefinitely, are wringable, dustless, unaffected by temperature, flame resistant, and impervious to rot and mildew. Booms have a strong mesh netting with metal clips for chaining together and easy retrieval from water. Disposal is easy™each sorbent incinerates to less than 0.2% ash.