This item is now discontinued. Please browse similar items
UltraTech 2130 Ultra DrainSeal, 36 x 36
Representative image only.
UltraTech Mfr # 2130 – Item # EW-86422-02
Prevent spills from entering drainage system
  • Temporarily "seals" to any smooth surface
  • Rugged, polyurethane construction resists tearing
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)36
  • Depth (in)3/8
  • Height (in)36
  • DescriptionUltra DrainSeal, 36 x 36
Key Features
  • Temporarily "seals" to any smooth surface
  • Rugged, polyurethane construction resists tearing
More About this Item
Drainseals feature a reinforcing mesh layer in between 2 layers of polyurethane for added strength and flexibility. Both sides quickly seal drains — no confusing "top" or "bottom".
Compliance: Helps comply with NPDES, 40 CFR 122.26 (1999)