UltraTech 2100 Ultra SpillBerm, 10 foot
Representative image only.
UltraTech Mfr # 2100 – Item # EW-86426-00
Flexible polyurethane conforms to cracks and crevices to seal flow
  • Ideal for sealing off drains or doorways during washdown applications
  • Non-absorbing for easy cleanup and reuse
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)120
  • Depth (in)2 1/4
  • Height (in)4
  • DescriptionUltra SpillBerm, 10 foot
Key Features
  • Ideal for sealing off drains or doorways during washdown applications
  • Non-absorbing for easy cleanup and reuse
More About this Item
Berms are flexible and easy to manipulate in the field – molded them into many shapes. Sold in 10 foot sections – order optional connector (sold separately below) to extend the length of your berm.
Compliance: Helps comply with NPDES, 40 CFR 122.26 (1999)