This item is now discontinued. Please browse similar items
UltraTech 9260 Ultra Passive Skimmer
Representative image only.
UltraTech Mfr # 9260 – Item # EW-86426-10
Absorb surface oil from catch basins and storm drains
  • One skimmer holds up to 2 gallons of oil
  • Compact and easy to use
SPECS & DESCRIPTION
Specifications & Description
  • Absorption4 gallons
  • Width (in)22
  • Depth (in)2
  • Height (in)14
  • DescriptionUltra Passive Skimmer
Key Features
  • One skimmer holds up to 2 gallons of oil
  • Compact and easy to use
More About this Item
Skimmer is constructed of a unique polymer absorbent – oil bonds to the polymer matrix preventing leaching. Will not create ponding or clog due to sediment. Economical skimmer floats on the water surface and continuously absorbs oil until it reaches maximum capacity.
Compliance: Helps comply with NPDES, 40 CFR 122.26 (1999)