This item is now discontinued. We recommend the following as suggested replacements: EW-78901-46
3M 11329-00000-20 Safety Eyewear, Clear frame, Clear lens, Anti-fog
Representative image only.
3M Mfr # 11329-00000-20 – Item # EW-86472-74
Lightweight design, feels almost like its not there
  • Sleek, 9.5-base lens provides superior protection while offering unobstructed view
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorClear
  • Lens ColorClear
  • Lens CoatingAntifog
  • ComplianceANSI Z87.1-2003
  • DescriptionSafety Eyewear, Clear frame, Clear lens, Anti-fog
Key Features
  • Sleek, 9.5-base lens provides superior protection while offering unobstructed view
More About this Item
Tough polycarbonate construction is designed to resist impacts and block 99.9% of UV rays. Features contoured, flexible temples, and universal-fit nosebridge.
Features an antifog coating.
Compliance: ANSI Z87.1-2003