Fisher Chemical C607SK4 Chloroform, Approx. 50ppm Pentene as Preservative/HPLC, Safe-Cote™ Amber Glass; 4 L

Representative image only.
Fisher Chemical Mfr # C607SK4 – Item # EW-88069-91
Quality and consistency your lab can count on

PRODUCT OPTIONS

$547.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • Empirical FormulaCHCl3
  • Linear FormulaCHCl3
  • ClassReagent Solvents
  • ChemistrySolutions
  • FormLiquid
  • SynonymsTrichloromethane
  • Volume (mL)4000
  • Formula Weight119.38
  • PackagingSafety Coated Amber Glass
  • DOT InformationChloroform; Class 6.1, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolHarmful
  • UN NumberUN1888
  • CAS Number67-66-3
  • DescriptionChloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 L)

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat