This item is now discontinued. Please browse similar items
Watlow CVC5HH01500500D On/Off Controller, J-Thermocouple Input (150 - 500F), Relay Output, 110V, Fahrenheit Only
Representative image only.
Watlow Mfr # CVC5HH01500500D – Item # EW-89002-12
An economical solution for applications requiring simple on/off control.
  • Fahrenheit or Celsius only operation
  • 1/8 DIN square panel mount unit with screw terminals
SPECS & DESCRIPTION
Specifications & Description
  • DIN Size0
  • Max Temperature (° F)1382
  • Min Temperature (° F)32
  • Resolution50,000 bits at full scale
  • Control Output 1relay
  • Display TypeSingle 4-digit LED displays, 0.3 in H
  • Power (VAC)110
  • DescriptionOn/Off Controller, J-Thermocouple Input (150 - 500F), Relay Output, 110V, Fahrenheit Only
Key Features
  • Fahrenheit or Celsius only operation
  • 1/8 DIN square panel mount unit with screw terminals
More About this Item
Watlow's agency-approved basic temperature controllers are compact and offer a cost effective control solution for less demanding applications requiring on/off control. These compact, 1/8-DIN square controllers have a microprocessor-designed platform — providing significant improvement in performance, repeatability and accuracy. Each product features a red, four-character, seven-segment LED display, set point selection, and definable operating temperature ranges. Controllers use a continuous-turn rotary selection switch and are designed to withstand harsh industrial environments.
Call our Applications Specialists for more information on these products or to special order a controller with other options.