This item is now discontinued. Please browse similar items
Temperature Transmitter RTD; 0 - 250F
Representative image only.
Item # EW-93500-12
SPECS & DESCRIPTION
Specifications & Description
  • Control Input RTD100 Ω
  • Output4 to 20 mA
  • Min Temperature (° F)0
  • Max Temperature (° F)250
  • Accuracy of Span±0.05%
  • Loop Power Supply11 to 40 VDC
  • DescriptionTemperature Transmitter RTD; 0 - 250F
More About this Item
Transmitter lets you increase the distance between your sensor and controller or panel meter. Linearized 4-20 mA ouput; nonisolated. Alpha coefficient is 0.00385 ohm/ohm/degC.
Use with three-wire, single-element RTDs—ideal for rigid-mounted probes with terminal block removed (order probes separately above). Transmitter may not be used with spring-loaded sheath probes. Screw terminal connections. RTD Transmitters let you increase the distance between your sensor and a controller or panel meter to 1000 ft. Can be used with two- or three-wire RTDs.