General Tools TI150 Digital Stopwatch/Clock with Flashing/Vibrating Alarm
Representative image only.
General Tools Mfr # TI150 – Item # EW-94411-16
  • 1 year warranty
$30.90USD / Each
$66.80 - $139.50USD / Each
for an additional
$66.80USD / Each
for an additional
$139.50USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity19 hours, 59 minutes or 19 minutes, 59 seconds
  • Timing ModesTimer switches between hour/min and min/sec, and has automatic memory
  • Count DirectionUp and down
  • Number of Channels1
  • Resolution1 sec, 1 min
  • AlarmThree different alarms—flashing red light, vibrating, and audible beep
  • DisplayLCD, 0.625\" H
  • Battery Type2 AAA (included)
  • Height (in)4
  • Outside Diameter (OD) (in)1 3/4
  • Height (cm)10.2
  • Outside Diameter (OD) (cm)4.5
  • DescriptionDigital Stopwatch/Clock with Flashing/Vibrating Alarm
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Timer features three different alarms—flashing red light, vibrating alarm, and audible beep. Timer switches between hour/min and min/sec, and has automatic memory. Timer can be worn on belt clip for convenience.