Jenco Economical 4 to 20 mA Transmitter; Type J; Operating Temperture; 14-158F/-10-70C
Representative image only.
Jenco Mfr # 77/02/+0000/+0300/CEL – Item # EW-94770-00
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionEconomical 4 to 20 mA Transmitter; Type J; Operating Temperture; 14-158F/-10-70C
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Economical 4 to 20 mA Transmitters have fixed 4 mA output at OdegC. Adjust span via three selectable solder jumper positions (units are shipped with jumper in position 2). Span accuracy is ±0.1%. Each transmitter features an LED that increases in brightness with increasing current output. Transmitters require a 12-36 VDC loop power supply. Screw terminal connections accept spade lugs or stripped-wire ends. Mounting holes are 33 mm apart.