Fowler 54-216-380 Digital Measuring Tape
Representative image only.
Fowler Mfr # 54-216-380 – Item # EW-97241-50
Read distance measurements with digital display
  • Freeze displayed measurements with Hold feature
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigital Measuring Tape
  • Warranty1 Year
1 Year Warranty
Key Features
  • Freeze displayed measurements with Hold feature
  • 1 year warranty
More About this Item
This digital tape measure is a fast and convenient tool for a wide range of length measurement applications. Measure up to 18 feet with one inch wide steel blade, and verify lengths on large LCD. The zero set button allows the origin point to be set at any time, eliminating the need for manually calculating comparison measurements. Display readings in feet, inches, fractions or millimeters. Five minute auto-off function conserves battery life.
What’s included: one lithium CR2032 battery.