This item is now discontinued. We recommend the following as suggested replacements: EW-98676-19
VALVE CHECK BRASS 1"NPT(F)
Representative image only.
Item # EW-98676-08
SPECS & DESCRIPTION
Specifications & Description
More About this Item
Operates efficiently in silence, with all of the functions of a swing check or vertical lift check valve. These are not suggested for installation in sewage ejector piping. Brass valves can be installed in any position.