This item is now discontinued. Please browse similar items
Thermo Scientific Haake 390-0100 Medium-High Range Rotational Viscometer Plus with RS232 interface 110-240V/ 50-60 Hz
Representative image only.
Thermo Scientific Haake Mfr # 390-0100 – Item # EW-98965-15
Accurately determines viscosity according to ISO 2555 and numerous ASTM standards
  • Visual and audio signals alert you to critical measuring conditions
  • Precisely control rotation with 21 pre-set speeds
  • Digital calibration of the torque with nationally traceable standards
SPECS & DESCRIPTION
Specifications & Description
  • Min Viscosity Range (cP)20
  • Max Viscosity Range (cP)40000000
  • Resolution0.1% full-scale
  • Repeatability±0.2% full-scale
  • Accuracy±1% full-scale
  • Rpm0.1, 0.2, 0.3, 0.5, 0.6, 1, 1.5, 4, 5, 6, 10, 12, 20, 30, 50, 60, 100, 200
  • Power (VAC)110 to 240
  • Power (Hz)50/60
  • DescriptionMedium-High Range Rotational Viscometer Plus with RS232 interface 110-240V/ 50-60 Hz
Includes
stand, set of spindles, 5-ft power cord.
Key Features
  • Visual and audio signals alert you to critical measuring conditions
  • Precisely control rotation with 21 pre-set speeds
  • Digital calibration of the torque with nationally traceable standards
More About this Item
These Thermo Scientific HAAKE Rotational 6 and 7 Plus viscometers are equipped for quality control, industrial, and laboratory viscosity tests, along with comparative measurements according to recognized standards. This viscometer measures the resistance of a test substance against a preset speed. These rotational viscometers have 21 speeds with built-in digital accuracy (±0.5% of the absolute value) by means of stepped motor. Dynamic viscosity and upper viscosity limit measurement results are displayed in both centipoise (cp) and millipascal (mPas), as well as percent torque, speed in rpm, and number of selected spindles.
The Thermo Scientific HAAKE 7 Plus viscometers feature a bi-directional RS-232 interface to transfer data to your PC or printer. Software for measurement and data evaluation is also available; purchase separately. A 100 Ω Pt temperature sensor monitors and displays the sample temperature in °F (°C).