This item is now discontinued. View the direct replacement: EW-99159-12 or browse similar items
Thermo Scientific 1150-1000 Nalgene Storm Water Sampler Mounting Kit, Reusable
Representative image only.
Thermo Scientific Mfr # 1150-1000 – Item # EW-99159-10
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionNalgene Storm Water Sampler Mounting Kit, Reusable
More About this Item
Mounting kit consists of black UV stabilized polyethylene tube, 24" steel stake, stainless steel worm clamp and lanyard, nylon rivets, polyester screen, wire hanger, and carabiner-style strap. Order disposable, white sampling bottles seperately at right.