Thermo Scientific Barnstead D4642-33 4-Holder Water Purification System; 240V

Representative image only.
Thermo Scientific Barnstead Mfr # D4642-33 – Item # EW-99252-15
Produce ASTM Type I water Recirculation pump ensures water purity
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • ASTM Water GradeType I
  • Max Flow Rate (L/min)2
  • Max Flow Rate (L/hour)120
  • Max Flow Rate (GPH)26.4
  • Number of Holders4
  • Inlet Port1/4\" NPT(F)
  • SourceTap water
  • Feed Water Requirements (pressure)100
  • Feed Water Requirements (Temperature)0-49⁰C
  • Max Inlet Water Temperature (° F)120
  • Min Inlet Water Temperature (° F)40
  • Max Inlet Water Temperature (° C)49
  • Min Inlet Water Temperature (° C)4
  • Min Inlet Pressure (PSI)5
  • Max Inlet Pressure (PSI)100
  • Min Inlet Pressure (bar)0.3
  • Max Inlet Pressure (bar)6.9
  • Power (VAC)240
  • Height (in)28
  • Depth (in)7.5
  • Width (in)35
  • Height (cm)71.1
  • Depth (cm)19
  • Width (cm)88.9
  • Description4-Holder Water Purification System; 240V
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneCanadian Standards Association

Key Features

  • 1 year warranty

More About this Item

Thermo Scientific Barnstead E-pure™ systems meet or exceed ASTM, CAP, and NCCLS requirements for bacteria free, Type I reagent grade water. Inert wetted parts prevent contamination of purified water. A digital resistivity meter (included) indicates water purity. A built-in, whisper-quiet recirculation pump with pressure regulator ensures Type I reagent grade water on demand. Order low-water and low-pressure pump protectors separately.
Choose from wall mountable three- or four-holder purification systems. Order disposable cartridges (required) by searching our Replacement Cartridge section; upgrade your system by adding an optional Remote Dispenser.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat