This item is now discontinued. We recommend the following as suggested replacements: EW-99271-31
Thermo Scientific Barnstead Batch Feed System with RF/UV
Representative image only.
Thermo Scientific Barnstead Mfr # D7035 – Item # EW-99261-02
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Inlet Water Temperature (° F)104
  • Min Inlet Water Temperature (° F)40
  • Max Inlet Water Temperature (° C)40
  • Min Inlet Water Temperature (° C)4
  • Max Inlet Pressure (PSI)100
  • Max Inlet Pressure (bar)6.9
  • DescriptionBatch Feed System with RF/UV
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
RF System uses a reservoir feed system. Unit is portable and lightweight—provides ultrapure water where you need it! TOC is less than 5 ppb.