This item is now discontinued. We recommend the following as suggested replacements: EW-99271-20

Thermo Scientific Barnstead Ultrafiltration System

Representative image only.
Thermo Scientific Barnstead Mfr # D7411 – Item # EW-99261-20


Specifications & Description

  • Max Flow Rate (L/min)1.2
  • Max Inlet Water Temperature (° F)120
  • Min Inlet Water Temperature (° F)40
  • Max Inlet Water Temperature (° C)49
  • Min Inlet Water Temperature (° C)4
  • Min Inlet Pressure (PSI)5
  • Max Inlet Pressure (PSI)100
  • Min Inlet Pressure (bar)0.345
  • Max Inlet Pressure (bar)6.9
  • DescriptionUltrafiltration System

More About this Item

UF System includes an ultrafilter that removes all particles greater that 10,000 daltons; produces pyrogen-free water. TOC is less than 10 ppb.





Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat