This item is now discontinued. We recommend the following as suggested replacements: EW-99271-40

Thermo Scientific Barnstead UF/UV Water Purification System; 1.3 L/min, 120 VAC

Representative image only.
Thermo Scientific Barnstead Mfr # D8611 – Item # EW-99261-40


Specifications & Description

  • Max Flow Rate (L/min)1.2
  • Max Inlet Water Temperature (° F)120
  • Min Inlet Water Temperature (° F)40
  • Max Inlet Water Temperature (° C)49
  • Min Inlet Water Temperature (° C)4
  • Min Inlet Pressure (PSI)5
  • Max Inlet Pressure (PSI)100
  • DescriptionUF/UV Water Purification System; 1.3 L/min, 120 VAC

More About this Item

UF/UV System is ideal for cell culture and monoclonal antibody production — produces pyrogen-, Rnase-, and Dnase-free water; and bacteria levels <1 CFU/100mL. TOC is less than 3 ppb.





Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat