Elga UHQIIXX11503 PURELAB UHQ II Water Purification System
Representative image only.
Elga Mfr # UHQIIXX11503 – Item # EW-99266-00
Combines the most efficient water purification techniques into a single compact system
  • 0.05-µm filter module ensures a particulate-free water supply for analytical instruments
  • UV reactor cell reduces bacterial content to <1 colony forming unit/ml
  • Recirculation pump maintains optimal water quality and inhibits microbial growth
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity15 L/day
  • Max Flow Rate (L/min)0.75
  • Height (in)16
  • Depth (in)16
  • Width (in)9
  • DescriptionPURELAB UHQ II Water Purification System
  • Warranty1 year
1 Year Warranty
Key Features
  • 0.05-µm filter module ensures a particulate-free water supply for analytical instruments
  • UV reactor cell reduces bacterial content to <1 colony forming unit/ml
  • Recirculation pump maintains optimal water quality and inhibits microbial growth
  • 1 year warranty
More About this Item
These compact systems incorporate multiple water purification technologies to provide up to 15 liters (UHQ II) or 50-liters (UHQ PS), per eight-hour day, of 18.2MΩ-cm ASTM Type 1 water. Units feature a short wavelength UV photochemical reactor cell, and adsorption, deionization, and microfiltration cartridges. Model 99266-00 includes a reverse osmosis cartridge enabling system to operate from a tap water feed. Model 99266-05 must be supplied with pretreated feed water. Built-in water quality meter automatically compensates for water temperature and continuously displays readings in MΩ-cm. Both units feature an internal 4-liter tank and recirculation pump.