DescriptionNANOpure Water Purification System, Life Science
Warranty1 year
Key Features
Automatic calibration verification and optional NIST traceable calibration module, 99275-56, lets you calibrate the system in the field
Purity up to 18.2 MΩ-cm — exceeds ASTM, CAP, and NCCLS Type 1 requirements
Ensure the organic carbon content of your water meets your most stringent standards with TOC monitoring models
1 year warranty
More About this Item
Thermo Scientific Barnstead NANOpure™ reagent grade water purification systems are designed to support tissue culture, PCR, electrophoresis, atomic absorption (AA), ICP, ICP/MS, ion chromatography (IC), gas chromatography (GC), and HPLC. Standby mode recirculates water for 10 minutes every hour during inactive periods. Additional standard features include compensated and uncompensated purity measurement, volumetric or timed dispensing of ultrapure water, automatic sanitization cycle, 0.2 µm absolute final filtration, and integral pressure regulating valve.
Thermo Scientific Barnstead NANOpure Life Science. Meets the most rigorous molecular biology application demands including PCR, electrophoresis, and cell and tissue culture. Features the latest advances in ion exchange and filtration technologies, dual wavelength UV bulb, ultrafilter, automatic system sanitization, automatic and manual ultrafilter flushing. Produces up to 1.5 liters per minute of water containing <0.001 Eu/ml of pyrogens, TOC values < 3 ppb, resistivities up to 18.2 M Ω-cm, and no detectable RNase, DNase, and DNA.