This item is now discontinued. We recommend the following as suggested replacements: EW-99289-30
Thermo Scientific Barnstead RO Reverse Osmosis System, 24 L/hr
Representative image only.
Thermo Scientific Barnstead Mfr # D12661 – Item # EW-99279-20
  • Space-saving design lets you mount RO system on the wall or the benchtop
  • Complete system includes everything you need — operation is easy and fully automatic
  • Membrane protection system prevents scale build up on the membrane
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (L/hour)24
  • Inlet Port3/8" OD compression
  • Max Inlet Water Temperature (° F)95
  • Min Inlet Water Temperature (° F)34
  • Max Inlet Water Temperature (° C)35
  • Min Inlet Water Temperature (° C)1
  • DescriptionRO Reverse Osmosis System, 24 L/hr
Key Features
  • Space-saving design lets you mount RO system on the wall or the benchtop
  • Complete system includes everything you need — operation is easy and fully automatic
  • Membrane protection system prevents scale build up on the membrane
More About this Item
These systems purify water for general laboratory use or as a feed source for ultrapure systems such as the EASYpure® II and NANOpure™. The 12 and 24 L/hr models use a diaphragm pump for quiet, efficient operation; the 6 L/hr model must be connected to an in-house line pressure.
The built-in membrane protection system extends the life of the membrane. It includes a special cartridge that softens the water, and an automatic flush that removes scale build-up on the membrane.
The control panel operates and monitors the main functions. The nine LEDs monitor the status of a function (operation, standby, flush), or warn you of a problem (low membrane pressure, low product purity, replace membrane, replace carbon filter, and liquid level in the storage reservoir).
Order the