This item is now discontinued. Please browse similar items
Compact General Purpose Still, 2 L/hr, 110 VAC
Representative image only.
Item # EW-99290-00
Distills up to 4 liters/hr
Specifications & Description
  • Capacity4 L/hr
  • Height (in)17.375
  • Outside Diameter (OD) (in)8.25
  • DescriptionCompact General Purpose Still, 2 L/hr, 110 VAC
More About this Item
These general purpose stills are designed for simple, economical, and reliable distillation. Stainless steel boilers and borosilicate glass condensers ensure distillate purity. All models have a nickel-plated heater. The heater is protected by a thermostat. Stills include a 3' cord with plug.
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
2 Questions | 2 Answers
Displaying questions 1-2
  • Q:
    We have this distillate in the lab and it has not been used for sometime until our current distillate system was out of service. I tried to test it. There was a small level valve has been found loose and has been tightened.
    I started the first run. Every thing was fine at first but after distillate drops came out to the collector bottle, I had to shutdown the devise as there was water coming out from the connection between the boiler and the condenser; it looks like the pressure has built up very quickly inside the boiler. Also, I was not able to monitor the water level inside the boiler due to water vapor drops that condensed inside boiler glass cover.
    I would be grateful if you can help me find out what went wrong?
    Thank You,
    Asked on 6/1/2010 by Walid from Canada
    1 answer
      It's possible after sitting unused some of the seals have weakened, which may lead to the leaking you are seeing. Please contact our technical application specialists for additional assistance (contact information can be found on the product web page).
      Answered on 6/3/2010 by David from Tech Support
  • Q:
    in your description of this product it describe the distilled water output as 2L/hr, 4L/hr, and 6L/hr which is it?
    Also where would i be able to purchase a replacement element for this product?
    Asked on 3/24/2010 by Anonymous
    1 answer
      We apologize for the confusion. This is a 2 L/hr output.
      Any replacement parts can be purchased through us. Please contact us for help in obtaining those parts.
      Answered on 3/24/2010 by Jaime from Technical Support
Displaying questions 1-2
Do you have a question about this product? 