Monarch 6180-022 Smart Laser Sensor
Representative image only.
Shimpo Mfr # 6180-022 – Item # EW-08212-59
Optimize performance of your tachometer with these accessories
  • Choose from a wide range of accessories to customize your tachometer to any application
  • “Smart” auto gain provides best performance in picking up target reflection.
  • TTL pulse output signal inverter Switch
  • Manual sensitivity knob provides dynamic fine tuning of sensor response
  • On/Off or Recharge Switch
  • Power/Battery Condition Indicator
  • Class 3R visible Laser and Nosepiece
  • Signal/Pulse/RS232 Output DIN connector Port
  • External DC power or recharger Port
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSLS-115/230 Smart Laser Sensor with NIST Calibration Certificate
  • Warranty1 year
1 Year Warranty
Includes
115/230 AC universal charger, SLS-CA-BNC 8-ft DIN output cable with BNC plug, and 12 inches of reflective tape.
Key Features
  • Choose from a wide range of accessories to customize your tachometer to any application
  • “Smart” auto gain provides best performance in picking up target reflection.
  • TTL pulse output signal inverter Switch
  • Manual sensitivity knob provides dynamic fine tuning of sensor response
  • On/Off or Recharge Switch
  • Power/Battery Condition Indicator
  • Class 3R visible Laser and Nosepiece
  • Signal/Pulse/RS232 Output DIN connector Port
  • External DC power or recharger Port
  • 1 year warranty
More About this Item
Keep your tachometer running at its best.
Smart Laser Sensor is an interal battery-powered optical sensor utilizing a visible Class 3R laser for a TTL pulse output. Operating range up to 65-ft (19.8m) with reflective tape and up to 3-ft (1m) off contrasting color targets, keyways, bolt heads, or blades.