Monarch 6280-032 PalmStrobe x Portable Stroboscope Pulse Input Cable
Representative image only.
Monarch Mfr # 6280-032 – Item # EW-98806-96
Removable, rechargeable battery pack allows unlimited measuring power
  • Compact, lightweight housing allows for convenient one-handed operation
  • Four-quadrant tuner makes stop action and slow motion inspections easy
PRODUCT OPTIONS
$36.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPalmStrobe x Portable Stroboscope Pulse Input Cable
Includes
removable battery pack with universal 115/230 VAC recharger and NIST-traceable calibration report supplied by the manufacturer. Pack 53510-01 also includes spare battery and field holster. Kit 53510-02 also includes two spare lamps and latching carrying c
Key Features
  • Compact, lightweight housing allows for convenient one-handed operation
  • Four-quadrant tuner makes stop action and slow motion inspections easy
More About this Item
The PalmStrobe x portable stroboscope is the industrial standard for high intensity, multifunction xenon stroboscopes. Memory holds eight commonly used flash patterns and the last flash rate used - no need to recalculate flash rates each time. Memory is retained even when the battery is recharging - recharge battery in or out of strobe.
The 6-digit, backlit LCD display is easy to read in both well-lit or dim locations. Additional features include rapid x2 and ÷2 of current flash rate, TTL-compatible input/output, and low-battery indicator.