Monarch DC1250-D00 2ch Recorder Isolated 12/24VDC Power Input
Representative image only.
Monarch Mfr # DC1250-D00 – Item # EW-49005-85
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Chart TypeDigital
  • Min Temperature (° F)-150
  • Max Temperature (° C)4350
  • Min Temperature (° C)-101
  • Description2ch Recorder Isolated 12/24VDC Power Input
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The DataChart 1250 is a powerful, compact digital recorder with intuitive operation, flexible input configuration and large memory capability. Combining small package size with extreme flexibility make the DataChart 1250 a powerful, yet economical, digital data recording system.
FEATURES
  • 2 universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTD's as well as frequency and pulse inputs
  • 4 internal alarm set points
  • 2 alarm relay outputs and 1 digital control input
  • Maximum sample storage rate of 100 samples per second per channel
  • Compact Flash cards up to 2 Gigabyte size
  • REVIEWS