Monarch DC1250-U10 Recorder; Universal AC Adapter/Rechargeable
Representative image only.
Monarch Mfr # DC1250-U10 – Item # EW-49005-91
  • 1 year warranty
$906.50USD / Each
$272.00 - $342.00USD / Each
for an additional
$272.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Chart TypeDigital
  • Min Temperature (° F)-150
  • Max Temperature (° C)4350
  • Min Temperature (° C)-101
  • Power (VAC)100 to 240
  • DescriptionRecorder; Universal AC Adapter/Rechargeable
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The DataChart 1250 is a powerful, compact digital recorder with intuitive operation, flexible input configuration and large memory capability. Combining small package size with extreme flexibility make the DataChart 1250 a powerful, yet economical, digital data recording system.
FEATURES
  • 2 universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTD's as well as frequency and pulse inputs
  • 4 internal alarm set points
  • 2 alarm relay outputs and 1 digital control input
  • Maximum sample storage rate of 100 samples per second per channel
  • Compact Flash cards up to 2 Gigabyte size
  • REVIEWS