This item is now discontinued. We recommend the following as suggested replacements: EW-04770-60
Mono-Mold Slim-Line Stir Bar, 2"L x 5/16" dia
Representative image only.
Mono-Mold Item # EW-04775-60
Get complete mixing
  • Slimmer profile and larger length to diameter ratio
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)2
  • Length (cm)5
  • Diameter (in)0.3125
  • Diameter (cm)0.8
  • DescriptionSlim-Line Stir Bar, 2\"L x 5/16\" dia
  • Warranty2 year
Two year warranty
Key Features
  • Slimmer profile and larger length to diameter ratio
  • 2 year warranty
More About this Item
These PTFE stir bars feature a slimmer profile and larger length to diameter ratio.