Morphix 382005-50 Gas Monitoring Badges for Mercury, box of 50.
Representative image only.
Morphix Mfr # 382005-50 – Item # EW-86563-06
Immediate visual indication when a specific chemical hazard is present
  • Exclamation mark warns of the presence of targeted hazard
  • Low cost and disposable
  • Ideal for daily exposure screening
PRODUCT OPTIONS
$414.50USD / Box of 50
SPECS & DESCRIPTION
Specifications & Description
  • Gases MeasuredMercury
  • DescriptionGas Monitoring Badges for Mercury, box of 50.
Key Features
  • Exclamation mark warns of the presence of targeted hazard
  • Low cost and disposable
  • Ideal for daily exposure screening
More About this Item
These low-cost, disposable chemical detector badges allow you to perform daily exposure screening. Badges require minimal training to use and require no calibration, extra equipment, or analysis. Badges attach easily to shirts or labcoat collars with the optional badge clips.
***NOTE: Due to special temperature requirements, all products must ship via expedited carrier - UPS Blue, UPS Red, or Fed-X Priority. Additional shipping charges vary by carrier and shipment weight. Contact a Customer Service Representative if freight estimate is required.***