Motic Instruments 282B Digital Microscope Camera, 2/3" CCD, Peltier cooled
Representative image only.
Motic Instruments Mfr # 282B – Item # EW-03908-76
Capture high-quality images with this scientific-grade CCD camera
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Resolution2580 x 1944
  • Image Sensor2/3\" CCD
  • System Requirements1 GB RAM or better, Windows® XP or better, Mac® OSX
  • PowerUSB (camera) and VAC (cooling)
  • Power (VAC)110 / 240
  • DescriptionDigital Microscope Camera, 2/3\" CCD, Peltier cooled
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Internal 4-frame buffer provides faster and more consistent frame rates. On-board image processing allows you to apply white balance or color corrections before the image is sent to your computer. Capture live, still, time-lapse, and video images with the included Motic Images Advanced 3.2 software. Software features include measurement, editing, amalgamation, manual/single point segmentation, single image assembly from multiple images, and multi-language display. USB 2.0 output provides easy communication with your PC or laptop and quick plug-and-play setup.
Peltier-cooled models monitor the temperature and humidity inside the camera case to help eliminate condensation brought on by rapid cooling. Set a target temperature through the software to facilitate repeatability. Achieve cooling of up to 10°C below ambient, enhancing the sensitivity of the image sensor.