Motic Instruments Moticam 10 Digital Microscope Camera, 1/2.5" CMOS
Representative image only.
Motic Instruments Mfr # 1100600100871 – Item # EW-03906-16
Fast plug-and-play cameras relay images from your microscope to your computer or monitor
  • Supplied software measures specimens and logs each image with automatic reference number, date, and time
PRODUCT OPTIONS
$1,640.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Resolution10.0 MP
  • Eyepiece Diameter (mm)12
  • Image Sensor1/2.5" CMOS
  • OutputUSB
  • System RequirementsWindows® XP and higher, Mac OSX®
  • Cable(s)USB
  • Power5 VDC via USB
  • DescriptionDigital Microscope Camera, 1/2.5" CMOS
Includes
focusable lens, calibration slide, two variable-diameter eyepiece adapters, macro tube, software, and mini USB cable.
Key Features
  • Supplied software measures specimens and logs each image with automatic reference number, date, and time
More About this Item
Capture electronic images and videos by converting your compound or stereo microscope into a powerful digital imaging system. These affordable digital cameras capture still or video images and instantly transmit them for real-time full screen viewing. Timed-interval shots provide time lapse observation—save, manipulate, print, and send!
Rapid view algorithms let you display at a faster frame rate while still being able to capture images at the highest resolution. These cameras are also great for capturing images of objects too large for a microscope stage. The included macro viewing tube is a stand that sits on the bench and holds the camera above a specimen.
Moticam 10 features the highest resolution of 10 megapixels, giving you the ability to convert images into 300 dpi files—ideal for documentation purposes!
Call Us At1-847-549-7600
Live chat