Multiprobe Flowblock Sampling System, acrylic, 1 GPM max
Representative image only.
Item # EW-35434-90
Recommended technology used by the U.S. EPA
  • Test your sample with up to four probes simultaneously
  • Ports fit a variety of probe sizes for use with most meters
  • Quick-connect inlet and outlet fittings for easy connections and cleaning
$613.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic
  • Sample Volume40mL
  • Range100 to 3785mL/min
  • DescriptionMultiprobe Flowblock Sampling System, acrylic, 1 GPM max
Key Features
  • Test your sample with up to four probes simultaneously
  • Ports fit a variety of probe sizes for use with most meters
  • Quick-connect inlet and outlet fittings for easy connections and cleaning
More About this Item
Multiprobe flowblock can be used directly in-line with most ground water pumping systems, and operates at flow rates of 100 mL/min to 1 GPM. Acrylic flowblock accepts up to four probe types, allowing you to measure several parameters of a sample at the same time. Sensors sit in direct line of the sample stream and keep void volume at less than 40 mL, reducing stirring dependence of sensors. Flowblock is compatible with most makes and models of water quality instrumentation.