MultiWash III Plate Washer with 12-Port Coaxial Manifold

Representative image only.
MultiWash III Mfr # 820-05(MULTIWASH3) – Item # EW-13054-16
  • Quick-connect fittings allow unit to be fully operational in 15 minutes
  • Rinse-on-demand feature for complete system cleaning at the touch of a button
  • Can handle full range of assay types from the most gentle wash to the most aggressive ELISA

PRODUCT OPTIONS

$5,952.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Well Volume (mL)2
  • Well ShapeIndependent horizontal and vertical adjustments allow any shape well to be accomodated
  • Number of Channels (Manifold)12
  • Min Residual Volume (mL)<1%/well
  • Max Number of Wash Cycles10
  • Min Dispense Volume (µL)50
  • Accuracy±3% at 300 μL
  • Height (in)8 1/2
  • Width (in)14 1/2
  • Length (in)11
  • Height (mm)292
  • DescriptionPlate Washer with 12-Port Coaxial Manifold
Two year warranty

Includes

manifold, slip connectors, reservoir kit, two power cords (US standard and European), fuse, rinse strips, dust cover, and, quick-start guide. Order additional manifolds separately.

Key Features

  • Quick-connect fittings allow unit to be fully operational in 15 minutes
  • Rinse-on-demand feature for complete system cleaning at the touch of a button
  • Can handle full range of assay types from the most gentle wash to the most aggressive ELISA

More About this Item

These plate washers feature easy-to-program wash routines, bottom washing, sweeping crosswise aspiration, dispense only and aspirate only functions, and overflow washing for continual dispensing while aspirating off the top for a thorough wash. Use the plate washer to perform cell washing; agglutination tests; cytotoxicity tests; and ELISA, EIA, FIA, and LIA tests.

Minimum residual volume is ≤1 μL per well. Washer has seven programmable wash operations. Independent adjustments of horizontal and vertical needle positions allow any shape well to be accommodated; extra aspiration adjustment for both horizontal and vertical needle positions can be stored in individual programs. Wetted materials include glass, nylon, PTFE, stainless steel, polypropylene, Tygon®, Delrin®, polyethylene, and Santoprene®.

The washer performs a 300-µL three-cycle wash on a full 96-well plate in less than two minutes. You control the dispense pressure. By programming and storing routines, you can switch between seven different modes at the touch of a button. The washer also features a floating manifold that adjusts to match each plate and allows the washer to perform special test procedures. All units are CE-approved, ETL-listed, and certified with CAN/CSA Std C22.2, No. 1010.1.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat