MV-10-1044-02 Diaphragm Valve, PFA, 1/4" NPT(F), 29"Hg to 100 psi
Representative image only.
Mfr # MV-10-1044-02 – Item # EW-31101-05
$331.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/4" NPT(F)
  • Orifice (in)0.25
  • Orifice (mm)6.35
  • Cv0.47
  • Wetted MaterialsPFA
  • DescriptionDiaphragm Valve, PFA, 1/4" NPT(F), 29"Hg to 100 psi
More About this Item
These diaphragm valves are made of PFA and feature no exposed metal parts. Choose between a two-way, three-way or sample diaphragm valve with various temperature and pressure ranges. Two-way valve 31101-05 includes bracket and screws. Three-way and sample valves include screws for mounting hardware.