MV-10-1344-02 Three-Way Diaphragm Valve, PFA, 1/4" NPT(F), 27"Hg to 45 psi
Representative image only.
Mfr # MV-10-1344-02 – Item # EW-98403-08
$399.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.25
  • Outlet Port1/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/4" NPT(F)
  • Orifice (in)0.25
  • Orifice (mm)6.35
  • Cv0.95
  • Max Pressure (PSI)45
  • Max Temperature (° F)200
  • Max Temperature (° C)93
  • DescriptionThree-Way Diaphragm Valve, PFA, 1/4" NPT(F), 27"Hg to 45 psi
More About this Item
These diaphragm valves are made of PFA and feature no exposed metal parts. Choose between a two-way, three-way or sample diaphragm valve with various temperature and pressure ranges. Two-way valve 31101-05 includes bracket and screws. Three-way and sample valves include screws for mounting hardware.