Red Lion 508TX 8 Port Industrial Ethernet Switch Din- Rail Or Panel Mount
Representative image only.
Red Lion Mfr # 508TX – Item # EW-17783-04
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Ports8
  • Max Input Voltage (VDC)30
  • Min Input Voltage (VDC)10
  • Max Input Current (mA)380
  • Description8 Port Industrial Ethernet Switch Din- Rail Or Panel Mount
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The 508TX and 508FX2 Series Industrial Ethernet Switches offer outstanding performance and ease of use. They are ideally suited for connecting Ethernet enabled industrial and/or security equipment and can be optionally configured with advanced Ethernet communication management functions. The N-Tron switches require no setup unless the advanced port functions are utilized. The six and eight 10/100BaseTX ports are auto sensing and auto configuring. Each copper port is automatically negotiated for maximum speed and performance by default. The two fiber optic ports on the 508FX2 support full 200 Mb/s communications via 100BaseFX. Bi-color LED's are provided to display the link status, link speed and activity of each port as well as power on/off status.
FEATURES
Full IEEE 802.3Compliance
Six 10/100 BaseTX RJ-45 Ports
Two 100 BaseFX Ports, ST or SC
Extended Environmental Specifications
Auto sensing 10/100BaseTX, Duplex, and MDIX
Up to 2.6 GB/s Throughput
Rugged Industrial DIN-RAIL Enclosure
Redundant Power Inputs (10-30 VDC)