Nasco Sampling B01109WA Storage Rack for Whirl-Pak Bags, holds 15
Representative image only.
Nasco Sampling Mfr # B01109 – Item # EW-06499-92
Convenient—collect, store and ship samples for lab analysis
  • Double grid rack provides extra support preventing bags from slipping down in the rack
  • Electrostatically applied powder coating creates a durable finish allowing the racks to be autoclaved
  • Optional rack handle is available making it easy to carry a full rack in one hand
  • Stand alone racks can hold 15 bags
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeStorage Rack
  • DescriptionStorage Rack for Whirl-Pak Bags, holds 15
  • Warranty1 year
1 Year Warranty
Key Features
  • Double grid rack provides extra support preventing bags from slipping down in the rack
  • Electrostatically applied powder coating creates a durable finish allowing the racks to be autoclaved
  • Optional rack handle is available making it easy to carry a full rack in one hand
  • Stand alone racks can hold 15 bags
  • 1 year warranty
More About this Item
Tailor your water collection system with the following components. WHIRL-PAK® sodium thiosulfate bags 06499-90 contain a 10-mg sodium thiosulfate tablet to dechlorinate water and allow bacteria to grow. Simply flip bag around wire/tape and bend wire to seal. Bag has a white write-on label for easy identification of samples. Clip bag to handy water scoop 06499-91 to obtain samples in lakes or rivers. Rack 06499-92 holds up to 15 filled bags for transport. Mail your sample with mailing tube 06499-93.