Nasco Sampling B01366WA Swing Sampler, polyethylene & polyurethane, 8 to 24 ft depth

Representative image only.
Nasco Sampling Mfr # B01366 – Item # EW-99155-32
Sampler swings allowing collections from a variety of different angles – including 90°
  • Easily collects samples from horizontally flowing streams

PRODUCT OPTIONS

$231.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialFiberglass pole, polyurethane bottle holder
  • Sample Volume960 mL
  • Bottle TypePolyethylene
  • Cap ConfigurationVinyl-lined Polyethylene
  • Length (ft)24
  • Length (cm)731.5
  • DescriptionSwing Sampler, polyethylene & polyurethane, 8 to 24 ft depth

Key Features

  • Easily collects samples from horizontally flowing streams

More About this Item

Use the swing sampler to obtain liquids, powders, or even small solids. Includes one 960-mL bottle and fits almost any other laboratory sample bottles.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat