Nasco Sampling B1098WA COLIWASA Sampler, HDPE, 5 ft depth
Representative image only.
Nasco Sampling Mfr # B01098WA – Item # EW-99155-00
Sample multiphase wastes with a wide range of viscosity, corrosivity, volatility, and solids content.*
  • Manufactured of high-density polyethylene
  • Designed to meet EPA standards
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCOLIWASA Sampler, HDPE, 5 ft depth
  • Warranty1 year
1 Year Warranty
Key Features
  • Manufactured of high-density polyethylene
  • Designed to meet EPA standards
  • 1 year warranty
More About this Item
Sampler consists of a 5-ft x 3/4" ID sampling tube, closure system, and a closure-locking mechanism.
*Note: Not for use with ketones, nitrobenzene, dimethylformamide, mesityl oxide, and tetrahydrofuran.