Nasco Sampling SC7150 Storage Rack, 18 x 100 mL; 1 Rack/Each

Representative image only.
Nasco Sampling Mfr # SC7150 – Item # EW-53201-17
Optimize performance of your Environmental Express HotBlock Digestion Cups
  • Watch glasses to cover your cups during digestion
  • Transfer racks to avoid spillage
  • Caps to close cups

PRODUCT OPTIONS

$35.90USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPowder-coated wire
  • Compatible With100 mL digestion cups
  • Length (in)6.5
  • Height (in)3
  • Width (in)12.75
  • DescriptionHotBlock Storage Rack, 18 x 100 mL; 1 Rack/Each

Key Features

  • Watch glasses to cover your cups during digestion
  • Transfer racks to avoid spillage
  • Caps to close cups

More About this Item

Choose from a wide range of accessories and replacement parts for your digestion cups

The storage rack is made from plastic-coated wire and designed to hold the 100-mL digestion cups.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat