Digital Gas Flowmeter for Air/N2/O2/CO2, 500 sccm
Representative image only.
Item # EW-68438-15
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±1.0%
  • DescriptionDigital Gas Flowmeter for Air/N2/O2/CO2, 500 sccm
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Measures Air, N2, O2 and CO2
Measures true gas flow that is independent of temperature and pressure changes. The flow is displayed directly in Standard Cubic Centimeter per Minute (SCCM).