Newport 88106K/N Right-Angle Surface Probe 4 1/2" Probe
Representative image only.
Newport Mfr # 88106K/N – Item # EW-08877-29
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeK
  • Max Temperature (° F)220
  • Max Temperature (° C)105
  • Connector TypeMini ANSI
  • Sheath Length (in)60
  • DescriptionRight-Angle Surface Probe 4 1/2\" Probe
More About this Item
The NEWPORT 88000 series surface/insertion probes are supplied with retractable sensor cable, with 1 ft extending to 5 ft. These cables are rated to 105 °C (220 °F). Available in type K and E calibrations, for temps up to 760 °C. Can be used on stationary, moving or rotating surfaces. Other styles available.