Nickel/SS Lab Spatulas with 0.5" Spoon x 1.25" Flat Ends, Hayman Style; 3/Pk
Representative image only.
Item # EW-06369-09
$37.50USD / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)5 3/4
  • Overall Length (cm)14.61
  • Material304 Stainless Steel, Nickel
  • SterileYes
  • DescriptionNickel/SS Lab Spatulas with 0.5" Spoon x 1.25" Flat Ends, Hayman Style; 3/Pk
More About this Item
Nickel/stainless steel spatulas come in a variety of end styles. All spatulas have a glazed finish, except for models 06369-05, and -16 which have a mirror finish.
Call Us At1-847-549-7600
Live chat