NIOSH N99 Particulate Respirator, Metal Nose Seal; 20/box
Representative image only.
Item # EW-78901-26
NIOSH approved and CDC listed particulate respirator
  • Light weight, soft comfortable, latex-free material
  • Cushioning nose foam for comfort
  • Conforms to NOISH 42CFR84
PRODUCT OPTIONS
$21.50USD / Box of 20
BUY MORE & SAVE
  • 1-9$21.50USD / Box of 20
  • 10-19$19.35USD / Box of 20
  • 20+$18.28USD / Box of 20
SPECS & DESCRIPTION
Specifications & Description
  • RatingN-99
  • NIOSH ApprovalNOISH 42CFR84
  • ComplianceNIOSH approved, CDC listed
  • DescriptionNIOSH N99 Particulate Respirator, Metal Nose Seal
Key Features
  • Light weight, soft comfortable, latex-free material
  • Cushioning nose foam for comfort
  • Conforms to NOISH 42CFR84
More About this Item
NIOSH approved particulate respirators protect against solid and liquid aerosols that do not contain oil. It features a flexible metal nose clip seal with stapled straps. Optional exhalation valve keeps you cool.