Nor-Lake NSXF051WMW/0 Freezer, Undercounter -30C, 5 cu ft with Legs, 115V/60Hz
Representative image only.
Nor-Lake Mfr # NSXF051WMW/0 – Item # EW-44174-30
Maintain uniform temperature for your critical sample storage
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)23
  • Freezer Capacity (Liters)651
  • Min Temperature - Refrigerator (° C)1
  • Max Temperature - Refrigerator (° C)10
  • Max Temperature - Freezer (° C)-6
  • Number of Doors1
  • Number of Shelves3
  • Power (VAC)120
  • Power (Hz)60
  • Power (amps)15
  • DescriptionFreezer, Undercounter -30C, 5 cu ft with Legs, 115V/60Hz
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Heavy duty refrigeration system features adjustable temperature from -30 to -25°C. Constructed of white epoxy coated galvinized steel, the high density polyurethane foam insulation helps maintain the set temperature. Solid-door features a pull door handle, magnetic gasket, and lock. Automatic defrost eliminates downtime.