North by Honeywell 75SC Cartridge for Full-Face and Half-Mask Respirators, Defender Multi-purpose
Representative image only.
North by Honeywell Mfr # 75SCL – Item # EW-81606-50
$19.50USD / Pkg of 2
BUY MORE & SAVE
  • 1-17$19.50USD / Pkg of 2
  • 18+$17.55USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • RatingP100
  • NIOSH ApprovalCertain organic vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Fluoride, Ammonia/Methylamine, Hydrogen Sulfide (escape only), Formaldehyde
  • ComplianceNIOSH
  • ColorOrange
  • DescriptionCartridge for Full-Face and Half-Mask Respirators, Defender Multi-purpose
More About this Item
Stay safe with replacement cartridges.