North by Honeywell 770030L-12 Half-Mask Respirator, Large

Representative image only.
North by Honeywell Mfr # 770030L – Item # EW-81606-14
Wraparound lens provides over 200° field of vision
  • Superior comfort and fit
  • Contoured flange ensures proper seal

PRODUCT OPTIONS

$47.10USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • SizeLarge
  • ComplianceNIOSH
  • ColorBlack
  • DescriptionHalf-Mask Respirator, Large

Key Features

  • Superior comfort and fit
  • Contoured flange ensures proper seal

More About this Item

These half-mask respirators are made of soft, pliable, non-allergenic silicone offering excellent comfort and fit. Low-profile positioning on the face gives a wide field of vision and room for protective eyewear. Contoured, extended side sealing flanges and design of nose area provide a good seal for easy communication or other facial motions. Removable cradle-suspension straps offer a balanced, comfortable and secure fit.

Order respirator cartridges separately according to the demands of your work environment.
Approval: NIOSH

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat